missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-methylaniline, 99%

Catalog No. AAA1381809
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1381809 10 g
AAA1381818 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1381809 Supplier Thermo Scientific Chemicals Supplier No. A1381809

CAS: 583-68-6 | C7H8BrN | 186.052 g/mol

The N-(3, 5-Dichlorophenyl) naphthaldimine was obtained from the reaction of 2-hy- droxynaphthalene-l-carbaldehyde (0.01 mol) with a solution of 2-bromo-4-methylaniline (0.01 mol) in 40 ml of ethanol. Reaction of 2-bromo-4-methylaniline was quite clean, albeit 3,4-diaminotoluene was isolated just in 37% yield after flash chromatography on silica gel. The bromomethylquinoline starting material 16 was synthesized in two steps using an optimized process route by condensing 2-bromo-4-methylaniline.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The N-(3, 5-Dichlorophenyl) naphthaldimine was obtained from the reaction of 2-hy- droxynaphthalene-l-carbaldehyde (0.01 mol) with a solution of 2-bromo-4-methylaniline (0.01 mol) in 40 ml of ethanol. Reaction of 2-bromo-4-methylaniline was quite clean, albeit 3,4-diaminotoluene was isolated just in 37% yield after flash chromatography on silica gel. The bromomethylquinoline starting material 16 was synthesized in two steps using an optimized process route by condensing 2-bromo-4-methylaniline.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 583-68-6
Molecular Formula C7H8BrN
Molecular Weight (g/mol) 186.052
MDL Number MFCD00007635
InChI Key UVRRJILIXQAAFK-UHFFFAOYSA-N
Synonym 2-bromo-p-toluidine, benzenamine, 2-bromo-4-methyl, 4-amino-3-bromotoluene, 2-bromo-4-methylbenzenamine, p-toluidine, 2-bromo, 2-bromo-4-methyl-phenylamine, 3-bromo-4-aminotoluene, 4-methyl-2-bromoaniline, 2-bromo-4-methyl aniline, 2-bromo-4-methyl-aniline
PubChem CID 11422
IUPAC Name 2-bromo-4-methylaniline
SMILES CC1=CC(=C(C=C1)N)Br

Specifications

Melting Point 14°C to 16°C
Density 1.494
Boiling Point 240°C to 242°C
Flash Point >110°C (230°F)
Odor Characteristic
Refractive Index 1.603
Quantity 10 g
UN Number UN2810
Beilstein 1931711
Solubility Information Insoluble in water.
Formula Weight 186.06
Percent Purity 99%
Chemical Name or Material 2-Bromo-4-methylaniline
Show More Show Less
Hazard Category H301+H311+H331-H315-H319-H335-H373
Hazard Statement GHS H Statement
H301-H311-H315-H319
Toxic if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P260-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P314-P330-P332+P313-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 209-515-7
RTECSNumber XU4300000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only