missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4'-methylacetophenone, 98%

Catalog No. p-7028089
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2521706 5 g
AAB2521714 25 g
AAB2521722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2521706 Supplier Thermo Scientific Chemicals Supplier No. B2521706
Only null left

CAS: 619-41-0 | C9H9BrO | 213.074 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 619-41-0
Molecular Formula C9H9BrO
Molecular Weight (g/mol) 213.074
MDL Number MFCD00000203
InChI Key KRVGXFREOJHJAX-UHFFFAOYSA-N
Synonym 2-bromo-4'-methylacetophenone, 2-bromo-1-p-tolyl ethanone, 4-methylphenacyl bromide, 2-bromo-1-4-methylphenyl ethanone, 2-bromo-1-p-tolylethanone, 2-bromo-4-methylacetophenone, 2-bromo-1-4-methylphenyl ethan-1-one, acetophenone, 2-bromo-4'-methyl, ethanone, 2-bromo-1-4-methylphenyl, p-methylphenacyl bromide
PubChem CID 69272
IUPAC Name 2-bromo-1-(4-methylphenyl)ethanone
SMILES CC1=CC=C(C=C1)C(=O)CBr

Specifications

Melting Point 50°C to 54°C
Boiling Point 105°C (0.1 mmHg)
Flash Point >110°C (230°F)
Quantity 5 g
Beilstein 607041
Formula Weight 213.08
Percent Purity 98%
Chemical Name or Material 2-Bromo-4'-methylacetophenone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 210-595-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only