accessibility menu, dialog, popup

UserName

CH3OBr

CAS RN 619-41-0

IUPAC Name: 2-bromo-1-(4-methylphenyl)ethan-1-one
Synonyms: 2-bromo-1-(4-methylphenyl)ethanone
Molecular Weight (g/mol): 213.07
Molecular Formula: C9H9BrO
InChi Key: KRVGXFREOJHJAX-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)C(=O)CBr
Molecular Weight (g/mol) 
  • (3)
  • (5)
Percent Purity 
  • (2)
  • (3)
  • (3)
Quantity 
  • (2)
  • (3)
  • (3)
Boiling Point 
  • (3)
  • (2)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (3)
  • (5)
  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)