missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-methoxybenzaldehyde, 98%

Catalog No. AAH61379MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH61379MD 250 mg
AAH6137903 1 g
AAH6137906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH61379MD Supplier Thermo Scientific Chemicals Supplier No. H61379MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 43192-31-0 | C8H7BrO2 | 215.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 43192-31-0
Molecular Formula C8H7BrO2
Molecular Weight (g/mol) 215.05
MDL Number MFCD09788636
InChI Key ODISAUHBLBVQKC-UHFFFAOYSA-N
Synonym benzaldehyde, 2-bromo-4-methoxy, benzaldehyde,2-bromo-4-methoxy, 3-bromo-4-formylanisole, 2-bromo-p-anisaldehyde, pubchem12556, acmc-209jtd, ksc232o5d, 2-bromanyl-4-methoxy-benzaldehyde
PubChem CID 11127641
IUPAC Name 2-bromo-4-methoxybenzaldehyde
SMILES COC1=CC(Br)=C(C=O)C=C1

Specifications

Quantity 250 mg
Formula Weight 215.04
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 2-Bromo-4-methoxybenzaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only