Learn More
2-Bromo-4'-methoxyacetophenone, 98%
CAS: 2632-13-5 | C9H9BrO2 | 229.07 g/mol
Supplier: Thermo Scientific Chemicals A1341514
Description
2-Bromo-4'-methoxyacetophenone, is used as a cell-permeable, covalent and potent protein tyrosine phosphatase inhibitor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Bromo-4′-methoxyacetophenone, is used as a cell-permeable, covalent and potent protein tyrosine phosphatase inhibitor.
Solubility
It is soluble in DMSO, water (partly miscible), most organic solvents, and methanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C
Specifications
| 2-Bromo-4'-methoxyacetophenone | |
| 68°C to 72°C | |
| C9H9BrO2 | |
| 25 g | |
| 743112 | |
| It is soluble in DMSO,water (partly miscible),most organic solvents,and methanol. | |
| COC1=CC=C(C=C1)C(=O)CBr | |
| 229.07 | |
| 229.08 |
| 2632-13-5 | |
| Irritating | |
| MFCD00000201 | |
| UN3261 | |
| 2-bromo-4'-methoxyacetophenone, 2-bromo-1-4-methoxyphenyl ethanone, ethanone, 2-bromo-1-4-methoxyphenyl, p-methoxyphenacyl bromide, 4'-methoxyphenacyl bromide, 2-bromo-1-4-methoxyphenyl ethan-1-one, 4-methoxyphenacyl bromide, bromomethyl 4-methoxyphenyl ketone, alpha-bromo-4-methoxyacetophenone, 2-bromo-4-methoxyacetophenone | |
| XQJAHBHCLXUGEP-UHFFFAOYSA-N | |
| 2-bromo-1-(4-methoxyphenyl)ethanone | |
| 4965 | |
| 98% |
Chemical Identifiers
| 2632-13-5 | |
| 229.07 | |
| XQJAHBHCLXUGEP-UHFFFAOYSA-N | |
| 4965 | |
| COC1=CC=C(C=C1)C(=O)CBr |
| C9H9BrO2 | |
| MFCD00000201 | |
| 2-bromo-4'-methoxyacetophenone, 2-bromo-1-4-methoxyphenyl ethanone, ethanone, 2-bromo-1-4-methoxyphenyl, p-methoxyphenacyl bromide, 4'-methoxyphenacyl bromide, 2-bromo-1-4-methoxyphenyl ethan-1-one, 4-methoxyphenacyl bromide, bromomethyl 4-methoxyphenyl ketone, alpha-bromo-4-methoxyacetophenone, 2-bromo-4-methoxyacetophenone | |
| 2-bromo-1-(4-methoxyphenyl)ethanone |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302-H314
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 220-118-8
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only