missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-methoxy-6-nitrophenol 98.0+%, TCI America™

Catalog No. B47231G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B47231G 1 g
B47235G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B47231G Supplier TCI America Supplier No. B47231G

Chemical Identifiers

CAS 115929-59-4
Molecular Formula C7H6BrNO4
Molecular Weight (g/mol) 248.032
MDL Number MFCD06797984
InChI Key GAVVLRJHVWIDPK-UHFFFAOYSA-N
PubChem CID 11265140
IUPAC Name 2-bromo-4-methoxy-6-nitrophenol
SMILES COC1=CC(=C(C(=C1)[N+](=O)[O-])O)Br

Specifications

Melting Point 118°C
Quantity 1 g
Formula Weight 248.03
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-4-methoxy-6-nitrophenol