missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-isopropylaniline 98.0+%, TCI America™

Catalog No. B47245G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B47245G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B47245G Supplier TCI America Supplier No. B47245G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 51605-97-1
Molecular Formula C9H12BrN
Molecular Weight (g/mol) 214.11
MDL Number MFCD00051585
InChI Key WEMDUNBELVTSRP-UHFFFAOYSA-N
Synonym 2-bromo-4-isopropylaniline, 2-bromo-4-isopropyl-phenylamine, 4-amino-2-bromocumene, benzenamine, 2-bromo-4-1-methylethyl, zlchem 981, pubchem17446, acmc-1anzo, 2-bromo-4-isopropyl aniline, 2-bromo4-isopropyl-phenylamine, 2-bromo-4-2-propyl aniline
PubChem CID 521337
IUPAC Name 2-bromo-4-(propan-2-yl)aniline
SMILES CC(C)C1=CC=C(N)C(Br)=C1

Specifications

Color Yellow
Boiling Point 111°C
Quantity 5 g
Formula Weight 214.11
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Bromo-4-isopropylaniline