missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-fluorotoluene, 98+%

Catalog No. AAA1279206
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1279206 5 g
AAA1279214 25 g
AAA1279222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1279206 Supplier Thermo Scientific Chemicals Supplier No. A1279206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1422-53-3 | C7H6BrF | 189.027 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1422-53-3
Molecular Formula C7H6BrF
Molecular Weight (g/mol) 189.027
MDL Number MFCD00040828
InChI Key SFGFOJPGCOYQJK-UHFFFAOYSA-N
Synonym 2-bromo-4-fluorotoluene, 4-fluoro-2-bromotoluene, 2-bromo-4-fluoro toluene, benzene, 2-bromo-4-fluoro-1-methyl, 2-bromo-4-fluoro-1-methyl-benzene, 2-brom-4-fluor-1-methylbenzol, pubchem1602, 2-bromo4-fluorotoluene, acmc-209cny, 2-bromo-4-fluoro-toluene
PubChem CID 74016
IUPAC Name 2-bromo-4-fluoro-1-methylbenzene
SMILES CC1=C(C=C(C=C1)F)Br

Specifications

Density 1.526
Boiling Point 170°C to 172°C
Flash Point 64°C (147°F)
Refractive Index 1.528
Quantity 5 g
Beilstein 1859038
Formula Weight 189.03
Percent Purity ≥98%
Chemical Name or Material 2-Bromo-4-fluorotoluene
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 215-830-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only