missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-4-fluorothiophene 95.0+%, TCI America™
Supplier: TCI America B36801G
Specifications
| 2-Bromo-4-fluorothiophene | |
| Yellow | |
| C4H2BrFS | |
| 1993 | |
| C1=C(SC=C1F)Br | |
| 181.022 | |
| 181.02 | |
| Liquid |
| 32431-65-5 | |
| 60°C | |
| 1 g | |
| UWXVZGDRWPSION-UHFFFAOYSA-N | |
| 2-bromo-4-fluorothiophene | |
| 14786520 | |
| ≥95.0% (GC) |
Chemical Identifiers
| 32431-65-5 | |
| 181.022 | |
| 14786520 | |
| C1=C(SC=C1F)Br |
| C4H2BrFS | |
| UWXVZGDRWPSION-UHFFFAOYSA-N | |
| 2-bromo-4-fluorothiophene |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator