accessibility menu, dialog, popup

UserName

SFBr

CAS RN 32431-65-5

IUPAC Name: 2-bromo-4-fluorothiophene
Synonyms: 2-bromo-4-fluorothiophene
Molecular Weight (g/mol): 181.02
Molecular Formula: C4H2BrFS
InChi Key: UWXVZGDRWPSION-UHFFFAOYSA-N
SMILES: FC1=CSC(Br)=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results