missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-fluorothiophene 95.0+%, TCI America™

Catalog No. B3680200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B3680200MG 200 mg
B36801G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B3680200MG Supplier TCI America Supplier No. B3680200MG
Only null left

Chemical Identifiers

CAS 32431-65-5
Molecular Formula C4H2BrFS
Molecular Weight (g/mol) 181.022
InChI Key UWXVZGDRWPSION-UHFFFAOYSA-N
PubChem CID 14786520
IUPAC Name 2-bromo-4-fluorothiophene
SMILES C1=C(SC=C1F)Br

Specifications

Color Yellow
Boiling Point 60°C
Quantity 200 mg
UN Number 1993
Formula Weight 181.02
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Bromo-4-fluorothiophene
TSCA No
Recommended Storage Refrigerator