missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-fluoropyridine 98.0+%, TCI America™

Catalog No. p-7123731
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B47601G 1 g
B47605G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B47601G Supplier TCI America Supplier No. B47601G
Only null left

Chemical Identifiers

CAS 357927-50-5
Molecular Formula C5H3BrFN
Molecular Weight (g/mol) 175.988
MDL Number MFCD09260909
InChI Key AIEATTRZKVGMBO-UHFFFAOYSA-N
Synonym 2-bromo-4-fluoro-pyridine, pyridine, 2-bromo-4-fluoro, 4-fluoro-2-bromopyridine, pubchem3004, acmc-209ij7, pybr71, ksc493m3n, 2-bromo-4-fluoropyridine, abbypharma ap-17-5103
PubChem CID 21885884
IUPAC Name 2-bromo-4-fluoropyridine
SMILES C1=CN=C(C=C1F)Br

Specifications

Color Yellow
Quantity 1 g
Formula Weight 175.99
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Bromo-4-fluoropyridine