accessibility menu, dialog, popup

UserName

NFBr

CAS RN 357927-50-5

IUPAC Name: 2-bromo-4-fluoropyridine
Synonyms: 2-bromo-4-fluoropyridine
Molecular Weight (g/mol): 175.99
Molecular Formula: C5H3BrFN
InChi Key: AIEATTRZKVGMBO-UHFFFAOYSA-N
SMILES: FC1=CC(Br)=NC=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)