missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-fluorophenol 98.0+%, TCI America™

Catalog No. p-7119116
Change view
Click to view available options
Quantity:
25 g
5 g
Catalog No. Quantity
B15555G 5 g
B155525G 25 g
2 options

Catalog No. B15555G

Supplier: TCI America B15555G

Only null left

Chemical Identifiers

CAS 496-69-5
Molecular Formula C6H4BrFO
Molecular Weight (g/mol) 190.999
MDL Number MFCD00010614
InChI Key MEYRABVEYCFHHB-UHFFFAOYSA-N
Synonym 2-bromo-4-fluoro-phenol, phenol, 2-bromo-4-fluoro, 4-fluoro-2-bromo phenol, pubchem1485, 2-bromo-4-flourophenol, 2-bromo4-fluoro-phenol, acmc-1artu, 2-bromo-4-fluoro phenol, ksc490k9b, akos akm01586
PubChem CID 605472
IUPAC Name 2-bromo-4-fluorophenol
SMILES C1=CC(=C(C=C1F)Br)O

Specifications

Melting Point 43°C
Boiling Point 89°C
Quantity 5 g
Formula Weight 191.00
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-4-fluorophenol