missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-fluorobenzotrifluoride 98.0+%, TCI America™

Catalog No. B33975G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B33975G 5 g
B339725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B33975G Supplier TCI America Supplier No. B33975G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 351003-21-9
Molecular Formula C7H3BrF4
Molecular Weight (g/mol) 243.00
MDL Number MFCD03094041
InChI Key XBBGYSIEJHXPLL-UHFFFAOYSA-N
Synonym 2-bromo-4-fluorobenzotrifluoride, 2-bromo-4-fluoro-1-trifluoromethyl benzene, 1-bromo-2-trifluoromethyl-5-fluorobenzene, benzene, 2-bromo-4-fluoro-1-trifluoromethyl, 2-bromo-4-florobenzotrifluoride, 2-bromo-4-fluoro-1-trifluoromethyl-benzene, pubchem1636, acmc-209icx, ksc911k1l, 2-bromo-4-fluorobenzo trifluoride
PubChem CID 2734909
IUPAC Name 2-bromo-4-fluoro-1-(trifluoromethyl)benzene
SMILES FC1=CC(Br)=C(C=C1)C(F)(F)F

Specifications

Color Yellow
Boiling Point 162°C
Quantity 5 g
Formula Weight 243.00
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Bromo-4-fluorobenzotrifluoride