missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-fluorobenzonitrile 98.0+%, TCI America™

Catalog No. p-7119746
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B27445G 5 g
B274425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B27445G Supplier TCI America Supplier No. B27445G
Only null left

Chemical Identifiers

CAS 36282-26-5
Molecular Formula C7H3BrFN
Molecular Weight (g/mol) 200.01
MDL Number MFCD00672924
InChI Key MNNDREXLRLDWEY-UHFFFAOYSA-N
Synonym 4-fluoro-2-bromobenzonitrile, 2-bromo-4-fluoro-benzonitrile, 2-bromo-4-fluorobenzenecarbonitrile, 2-cyano-5-fluorobromobenzene, benzonitrile, 2-bromo-4-fluoro, pubchem3217, acmc-1ajip, ksc490q6h, 2-bromo-4-fluoro benzonitrile, 2-bromo-4-fluorobenzonitrile
PubChem CID 118939
IUPAC Name 2-bromo-4-fluorobenzonitrile
SMILES FC1=CC=C(C#N)C(Br)=C1

Specifications

Melting Point 77°C
Quantity 5 g
UN Number 3439
Formula Weight 200.01
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-4-fluorobenzonitrile