missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-fluorobenzoic acid, 98%

Catalog No. AAL1932114
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1932114 25 g
AAL1932106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1932114 Supplier Thermo Scientific Chemicals Supplier No. L1932114
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1006-41-3 | C7H4BrFO2 | 219.009 g/mol

2-Bromo-4-fluorobenzoic acid is used in the synthesis of 3-fluoro-8-(methylthio)dibenzo[b,f]thiepin-10(11H)-one, 2-fluoro-8-(methylthio)dibenzo[b,f]thiepin-10(11H)-one, 2-((2-carboxy-5-fluorophenyl)amino)-3-methoxybenzoic acid and 2-bromo-4-fluorobenzamide. It is also used in the preparation of boron-containing anti-fungal agents, in the treatment of onychomycosis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-4-fluorobenzoic acid is used in the synthesis of 3-fluoro-8-(methylthio)dibenzo[b,f]thiepin-10(11H)-one, 2-fluoro-8-(methylthio)dibenzo[b,f]thiepin-10(11H)-one, 2-((2-carboxy-5-fluorophenyl)amino)-3-methoxybenzoic acid and 2-bromo-4-fluorobenzamide. It is also used in the preparation of boron-containing anti-fungal agents, in the treatment of onychomycosis.

Solubility
Soluble in water and methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 1006-41-3
Molecular Formula C7H4BrFO2
Molecular Weight (g/mol) 219.009
MDL Number MFCD00055370
InChI Key RRKPMLZRLKTDQV-UHFFFAOYSA-N
Synonym 2-bromo-4-fluorobenzoicacid, 4-fluoro-2-bromobenzoic acid, 2-bromo-4-fluoro-benzoic acid, benzoic acid, 2-bromo-4-fluoro, rarechem al bo 1996, buttpark 18\02-85, 2-bromo-4-fluorbenzoic acid, pubchem1314, 2-bromo-4-fluorobenzoic, 2-bromofluorobenzoic acid
PubChem CID 1268169
IUPAC Name 2-bromo-4-fluorobenzoic acid
SMILES C1=CC(=C(C=C1F)Br)C(=O)O

Specifications

Melting Point 167°C to 169°C
Quantity 25 g
Beilstein 2614292
Solubility Information Soluble in water and methanol.
Formula Weight 219.01
Percent Purity 98%
Chemical Name or Material 2-Bromo-4-fluorobenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only