missing translation for 'onlineSavingsMsg'
Learn More

2'-Bromo-4'-fluoroacetophenone, 98%

CAS: 1006-39-9 | C8H6BrFO | 217.04 g/mol

$127.10 - $423.67

Chemical Identifiers

CAS 1006-39-9
Molecular Formula C8H6BrFO
Molecular Weight (g/mol) 217.04
MDL Number MFCD00077464
InChI Key RCXFSBRMWBFWMH-UHFFFAOYSA-N
Synonym 2'-bromo-4'-fluoroacetophenone, 1-2-bromo-4-fluorophenyl ethanone, 1-acetyl-2-bromo-4-fluorobenzene, 1-2-bromo-4-fluorophenyl ethan-1-one, ethanone, 1-2-bromo-4-fluorophenyl, 1-2-bromo-4-fluoro-phenyl ethanone, pubchem8533, acmc-2097rj, ksc499c4r, timtec-bb sbb006561
PubChem CID 2736304
IUPAC Name 1-(2-bromo-4-fluorophenyl)ethanone
SMILES CC(=O)C1=C(Br)C=C(F)C=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1932003
View Documents
Thermo Scientific Chemicals
L1932003
1 g
Each for $127.10
Only null left
 
AAL1932006
View Documents
Thermo Scientific Chemicals
L1932006
5 g
Each for $423.67
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1006-39-9
217.04
RCXFSBRMWBFWMH-UHFFFAOYSA-N
2736304
CC(=O)C1=C(Br)C=C(F)C=C1
C8H6BrFO
MFCD00077464
2'-bromo-4'-fluoroacetophenone, 1-2-bromo-4-fluorophenyl ethanone, 1-acetyl-2-bromo-4-fluorobenzene, 1-2-bromo-4-fluorophenyl ethan-1-one, ethanone, 1-2-bromo-4-fluorophenyl, 1-2-bromo-4-fluoro-phenyl ethanone, pubchem8533, acmc-2097rj, ksc499c4r, timtec-bb sbb006561
1-(2-bromo-4-fluorophenyl)ethanone

Specifications

1006-39-9
>110°C (230°F)
1.549
1 g
2'-bromo-4'-fluoroacetophenone, 1-2-bromo-4-fluorophenyl ethanone, 1-acetyl-2-bromo-4-fluorobenzene, 1-2-bromo-4-fluorophenyl ethan-1-one, ethanone, 1-2-bromo-4-fluorophenyl, 1-2-bromo-4-fluoro-phenyl ethanone, pubchem8533, acmc-2097rj, ksc499c4r, timtec-bb sbb006561
CC(=O)C1=C(Br)C=C(F)C=C1
217.04
217.04
2'-Bromo-4'-fluoroacetophenone
150°C (12 mmHg)
C8H6BrFO
MFCD00077464
2249797
RCXFSBRMWBFWMH-UHFFFAOYSA-N
1-(2-bromo-4-fluorophenyl)ethanone
2736304
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only