CAS RN 1006-39-9
IUPAC Name:
1-(2-bromo-4-fluorophenyl)ethan-1-one
Synonyms:
1-(2-bromo-4-fluorophenyl)ethanone
Molecular Weight (g/mol):
217.04
Molecular Formula:
C8H6BrFO
InChi Key:
RCXFSBRMWBFWMH-UHFFFAOYSA-N
SMILES:
CC(=O)C1=C(Br)C=C(F)C=C1
1
–
1
of
1
results
1
Promotions Available
2'-Bromo-4'-fluoroacetophenone, 98%
2'-Bromo-4'-fluoroacetophenone, 98%
CAS: 1006-39-9 Molecular Formula: C8H6BrFO Molecular Weight (g/mol): 217.04 MDL Number: MFCD00077464 InChI Key: RCXFSBRMWBFWMH-UHFFFAOYSA-N Synonym: 2'-bromo-4'-fluoroacetophenone,1-2-bromo-4-fluorophenyl ethanone,1-acetyl-2-bromo-4-fluorobenzene,1-2-bromo-4-fluorophenyl ethan-1-one,ethanone, 1-2-bromo-4-fluorophenyl,1-2-bromo-4-fluoro-phenyl ethanone,pubchem8533,acmc-2097rj,ksc499c4r,timtec-bb sbb006561 PubChem CID: 2736304 IUPAC Name: 1-(2-bromo-4-fluorophenyl)ethanone SMILES: CC(=O)C1=C(Br)C=C(F)C=C1