missing translation for 'onlineSavingsMsg'
Learn More

2'-Bromo-4'-fluoroacetophenone, 98%

Catalog No. AAL1932003
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1932003 1 g
AAL1932006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1932003 Supplier Thermo Scientific Chemicals Supplier No. L1932003
Only null left

CAS: 1006-39-9 | C8H6BrFO | 217.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1006-39-9
Molecular Formula C8H6BrFO
Molecular Weight (g/mol) 217.04
MDL Number MFCD00077464
InChI Key RCXFSBRMWBFWMH-UHFFFAOYSA-N
Synonym 2'-bromo-4'-fluoroacetophenone, 1-2-bromo-4-fluorophenyl ethanone, 1-acetyl-2-bromo-4-fluorobenzene, 1-2-bromo-4-fluorophenyl ethan-1-one, ethanone, 1-2-bromo-4-fluorophenyl, 1-2-bromo-4-fluoro-phenyl ethanone, pubchem8533, acmc-2097rj, ksc499c4r, timtec-bb sbb006561
PubChem CID 2736304
IUPAC Name 1-(2-bromo-4-fluorophenyl)ethanone
SMILES CC(=O)C1=C(Br)C=C(F)C=C1

Specifications

Boiling Point 150°C (12 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.549
Quantity 1 g
Beilstein 2249797
Formula Weight 217.04
Percent Purity 98%
Chemical Name or Material 2'-Bromo-4'-fluoroacetophenone
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only