missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-4-fluoro-6-nitroaniline 98.0+%, TCI America™
Supplier: TCI America B48721G
Specifications
| 2-Bromo-4-fluoro-6-nitroaniline | |
| 77°C | |
| MFCD07779528 | |
| 2-bromo-4-fluoro-6-nitrobenzenamine, 2-bromo-4-fluoro-6-nitrophenylamine, benzenamine, 2-bromo-4-fluoro-6-nitro, 2-amino-3-bromo-5-fluoronitrobenzene, pubchem16316, ksc495m4t, 4-fluoro-2-bromo-6-nitroaniline, 4-fluoro-2-nitro-6-bromoaniline, 6-bromo-4-fluoro-2-nitroaniline, 2-bromo-4-fluoro-6-nitroaniline | |
| NC1=C(Br)C=C(F)C=C1[N+]([O-])=O | |
| 235.01 | |
| 235.01 | |
| Crystalline Powder |
| 10472-88-5 | |
| C6H4BrFN2O2 | |
| 1 g | |
| HCYDUPDSEDHSQB-UHFFFAOYSA-N | |
| 2-bromo-4-fluoro-6-nitroaniline | |
| 292004 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 10472-88-5 | |
| 235.01 | |
| HCYDUPDSEDHSQB-UHFFFAOYSA-N | |
| 292004 | |
| NC1=C(Br)C=C(F)C=C1[N+]([O-])=O |
| C6H4BrFN2O2 | |
| MFCD07779528 | |
| 2-bromo-4-fluoro-6-nitrobenzenamine, 2-bromo-4-fluoro-6-nitrophenylamine, benzenamine, 2-bromo-4-fluoro-6-nitro, 2-amino-3-bromo-5-fluoronitrobenzene, pubchem16316, ksc495m4t, 4-fluoro-2-bromo-6-nitroaniline, 4-fluoro-2-nitro-6-bromoaniline, 6-bromo-4-fluoro-2-nitroaniline, 2-bromo-4-fluoro-6-nitroaniline | |
| 2-bromo-4-fluoro-6-nitroaniline |
Safety and Handling
TSCA : No