missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-fluoro-1-iodobenzene 98.0+%, TCI America™

Catalog No. B30985G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B30985G 5 g
B30981G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B30985G Supplier TCI America Supplier No. B30985G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 202865-73-4
Molecular Formula C6H3BrFI
Molecular Weight (g/mol) 300.897
MDL Number MFCD00142868
InChI Key GKGPZAZTXZCUMU-UHFFFAOYSA-N
Synonym 2-bromo-4-fluoroiodobenzene, 2-bromo-4-fluoro-1-iodo-benzene, 2'-bromo-4-fluoro-1-iodobenzene, benzene, 2-bromo-4-fluoro-1-iodo, pubchem3234, acmc-1cll6, ksc495e7f, 2-bromo-4-fluoro-iodobenzene, 1-fluoro-3-bromo-4-iodobenzene
PubChem CID 2773372
IUPAC Name 2-bromo-4-fluoro-1-iodobenzene
SMILES C1=CC(=C(C=C1F)Br)I

Specifications

Color Yellow
Boiling Point 225°C
Quantity 5 g
Formula Weight 300.90
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Bromo-4-fluoro-1-iodobenzene