missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-fluoro-1-iodobenzene, 98+%

Catalog No. AAL2014403
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL2014403 1 g
AAL2014406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL2014403 Supplier Thermo Scientific Chemicals Supplier No. L2014403

CAS: 202865-73-4 | C6H3BrFI | 300.897 g/mol

It is employed in the synthesis of tetrasubstituted alkenes by stereo- and regioselective stannyllithiation of diarylacetylenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed in the synthesis of tetrasubstituted alkenes by stereo- and regioselective stannyllithiation of diarylacetylenes.

Solubility
Not miscible or difficult to mix in water.

Notes
Light sensitive. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store in dark.

Chemical Identifiers

CAS 202865-73-4
Molecular Formula C6H3BrFI
Molecular Weight (g/mol) 300.897
MDL Number MFCD00142868
InChI Key GKGPZAZTXZCUMU-UHFFFAOYSA-N
Synonym 2-bromo-4-fluoroiodobenzene, 2-bromo-4-fluoro-1-iodo-benzene, 2'-bromo-4-fluoro-1-iodobenzene, benzene, 2-bromo-4-fluoro-1-iodo, pubchem3234, acmc-1cll6, ksc495e7f, 2-bromo-4-fluoro-iodobenzene, 1-fluoro-3-bromo-4-iodobenzene
PubChem CID 2773372
IUPAC Name 2-bromo-4-fluoro-1-iodobenzene
SMILES C1=CC(=C(C=C1F)Br)I

Specifications

Density 1.33
Boiling Point 224°C to 225°C
Flash Point >110°C (230°F)
Refractive Index 1.629
Quantity 1 g
Beilstein 6288420
Sensitivity Light sensitive
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 300.9
Percent Purity ≥98%
Chemical Name or Material 2-Bromo-4-fluoro-1-iodobenzene
Show More Show Less
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only