missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-cyanopyridine, 97%

Catalog No. p-7029698
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2719103 1 g
AAH2719106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2719103 Supplier Thermo Scientific Chemicals Supplier No. H2719103
Only null left

CAS: 10386-27-3 | C6H3BrN2 | 183.01 g/mol

Mainly bromo compounds find application as dyes. Also in pharmaceutical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Mainly bromo compounds find application as dyes. Also in pharmaceutical industry.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 10386-27-3
Molecular Formula C6H3BrN2
Molecular Weight (g/mol) 183.01
MDL Number MFCD07367879
InChI Key AWSJFEKOXQBDSL-UHFFFAOYSA-N
Synonym 2-bromo-4-cyanopyridine, 2-bromoisonicotinonitrile, 2-bromo-isoniconinonitrile, 4-pyridinecarbonitrile, 2-bromo, 2-bromo-isonicotinonitrile, 2-bromo-4-pyridinecarbonitrile, pubchem14366, 2-bromoisonicotnonitrile, 2-bromo-4-cyano pyridine, 2-bromo-4-cyano-pyridine
PubChem CID 11735544
IUPAC Name 2-bromopyridine-4-carbonitrile
SMILES BrC1=NC=CC(=C1)C#N

Specifications

Melting Point 70°C to 74°C
Odor Odorless
Quantity 1 g
Solubility Information Insoluble in water.
Formula Weight 183.01
Percent Purity 97%
Chemical Name or Material 2-Bromo-4-cyanopyridine
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only