accessibility menu, dialog, popup

UserName

CAS RN 10386-27-3

IUPAC Name: 2-bromopyridine-4-carbonitrile
Synonyms: 2-bromopyridine-4-carbonitrile
Molecular Weight (g/mol): 183.01
Molecular Formula: C6H3BrN2
InChi Key: AWSJFEKOXQBDSL-UHFFFAOYSA-N
SMILES: BrC1=NC=CC(=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results