missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-4-chlorotoluene 98.0+%, TCI America™
Supplier: TCI America B303125G
Specifications
| 2-Bromo-4-chlorotoluene | |
| Yellow | |
| 25 g | |
| C7H6BrCl | |
| 2-bromo-4-chlorotoluene, benzene, 2-bromo-4-chloro-1-methyl, 4-chloro-2-bromotoluene, toluene, 2-bromo-4-chloro, p-chlor brom toluol, pubchem3599, acmc-209gvg, 2-bromo-4-chloro toluene, 2-bromo-4-chloro-toluene, ksc493k9b | |
| CC1=C(Br)C=C(Cl)C=C1 | |
| 205.48 | |
| 205.48 |
| 27139-97-5 | |
| 114°C | |
| Clear Liquid at 20°C | |
| MFCD00060650 | |
| CSUUXPHPCXHYGY-UHFFFAOYSA-N | |
| 2-bromo-4-chloro-1-methylbenzene | |
| 609898 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 27139-97-5 | |
| 205.48 | |
| CSUUXPHPCXHYGY-UHFFFAOYSA-N | |
| 609898 | |
| CC1=C(Br)C=C(Cl)C=C1 |
| C7H6BrCl | |
| MFCD00060650 | |
| 2-bromo-4-chlorotoluene, benzene, 2-bromo-4-chloro-1-methyl, 4-chloro-2-bromotoluene, toluene, 2-bromo-4-chloro, p-chlor brom toluol, pubchem3599, acmc-209gvg, 2-bromo-4-chloro toluene, 2-bromo-4-chloro-toluene, ksc493k9b | |
| 2-bromo-4-chloro-1-methylbenzene |
Safety and Handling
TSCA : No