missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-chlorotoluene 98.0+%, TCI America™

Catalog No. p-7119903
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B30315G 5 g
B303125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B30315G Supplier TCI America Supplier No. B30315G

Chemical Identifiers

CAS 27139-97-5
Molecular Formula C7H6BrCl
Molecular Weight (g/mol) 205.48
MDL Number MFCD00060650
InChI Key CSUUXPHPCXHYGY-UHFFFAOYSA-N
Synonym 2-bromo-4-chlorotoluene, benzene, 2-bromo-4-chloro-1-methyl, 4-chloro-2-bromotoluene, toluene, 2-bromo-4-chloro, p-chlor brom toluol, pubchem3599, acmc-209gvg, 2-bromo-4-chloro toluene, 2-bromo-4-chloro-toluene, ksc493k9b
PubChem CID 609898
IUPAC Name 2-bromo-4-chloro-1-methylbenzene
SMILES CC1=C(Br)C=C(Cl)C=C1

Specifications

Color Yellow
Quantity 5 g
Physical Form Clear Liquid at 20°C
Boiling Point 114°C
Formula Weight 205.48
Percent Purity ≥98.0% (GC)
Chemical Name or Material 2-Bromo-4-chlorotoluene