missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-chloroaniline, 98%

Catalog No. AAL0380814
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0380814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0380814 Supplier Thermo Scientific Chemicals Supplier No. L0380814
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 873-38-1 | C6H5BrClN | 206.467 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 873-38-1
Molecular Formula C6H5BrClN
Molecular Weight (g/mol) 206.467
MDL Number MFCD00041313
InChI Key SYTBIFURTZACKR-UHFFFAOYSA-N
Synonym benzenamine, 2-bromo-4-chloro, 1-amino-2-bromo-4-chlorobenzene, 2-bromo-4-chloro-phenylamine, 2-bromo-4-chloro aniline, 2-bromo-4-chlorophenylamine, pubchem3576, acmc-1bkdm, 2-bromo-4-chloro-aniline, 4-chloro-2-bromoaniline, 2-bromo-4-chloroaniline
PubChem CID 70110
IUPAC Name 2-bromo-4-chloroaniline
SMILES C1=CC(=C(C=C1Cl)Br)N

Specifications

Melting Point 63°C to 68°C
Boiling Point 127°C (20 mmHg)
Packaging Glass bottle
Quantity 25 g
UN Number UN2811
Beilstein 2802563
Formula Weight 206.47
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-4-chloroaniline
Hazard Category H301+H311+H331-H315-H319-H373
Precautionary Statement P260-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P314-P330-P332+P313-P361-P363-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 212-837-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only