missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-chloro-1-iodobenzene, 98%

Catalog No. AAB2540906
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2540906 5 g
AAB2540914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2540906 Supplier Thermo Scientific Chemicals Supplier No. B2540906
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 31928-44-6 | C6H3BrClI | 317.348 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 31928-44-6
Molecular Formula C6H3BrClI
Molecular Weight (g/mol) 317.348
MDL Number MFCD00672944
InChI Key CXHXFDQEFKFYQJ-UHFFFAOYSA-N
Synonym 2-bromo-4-chloroiodobenzene, benzene, 2-bromo-4-chloro-1-iodo, acmc-209hpo, ksc494q6f, 1-iodo-2-bromo-4-chlorobenzene, 2-bromo-1-iodo-4-chlorobenzene
PubChem CID 13741326
IUPAC Name 2-bromo-4-chloro-1-iodobenzene
SMILES C1=CC(=C(C=C1Cl)Br)I

Specifications

Quantity 5 g
Sensitivity Light sensitive
Formula Weight 317.36
Percent Purity 98%
Chemical Name or Material 2-Bromo-4-chloro-1-iodobenzene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only