missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-chloro-1-fluorobenzene 98.0+%, TCI America™

Catalog No. B29565G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B29565G 5 g
B295625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B29565G Supplier TCI America Supplier No. B29565G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1996-30-1
Molecular Formula C6H3BrClF
Molecular Weight (g/mol) 209.442
MDL Number MFCD00672934
InChI Key YFFUYGSLQXVHMB-UHFFFAOYSA-N
Synonym 3-bromo-4-fluorochlorobenzene, 3-bromo-1-chloro-4-fluorobenzene, 2-fluoro-5-chlorobromobenzene, 1-bromo-5-chloro-2-fluorobenzene, 1-bromo-3-chloro-6-fluorobenzene, 5-chloro-2-fluorobromobenzene, 2-bromo-4-chloro-1-fluoro-benzene, 2-bromo-4-chlorofluorobenzene, benzene, 2-bromo-4-chloro-1-fluoro, pubchem3210
PubChem CID 2773264
IUPAC Name 2-bromo-4-chloro-1-fluorobenzene
SMILES C1=CC(=C(C=C1Cl)Br)F

Specifications

Color Yellow
Boiling Point 90°C
Quantity 5 g
Formula Weight 209.44
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Bromo-4-chloro-1-fluorobenzene