missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-chloro-1-fluorobenzene, 97%

CAS: 1996-30-1 | C6H3BrClF | 209.442 g/mol

$144.75 - $451.92

Chemical Identifiers

CAS 1996-30-1
Molecular Formula C6H3BrClF
Molecular Weight (g/mol) 209.442
MDL Number MFCD00672934
InChI Key YFFUYGSLQXVHMB-UHFFFAOYSA-N
Synonym 3-bromo-4-fluorochlorobenzene, 3-bromo-1-chloro-4-fluorobenzene, 2-fluoro-5-chlorobromobenzene, 1-bromo-5-chloro-2-fluorobenzene, 1-bromo-3-chloro-6-fluorobenzene, 5-chloro-2-fluorobromobenzene, 2-bromo-4-chloro-1-fluoro-benzene, 2-bromo-4-chlorofluorobenzene, benzene, 2-bromo-4-chloro-1-fluoro, pubchem3210
PubChem CID 2773264
IUPAC Name 2-bromo-4-chloro-1-fluorobenzene
SMILES C1=CC(=C(C=C1Cl)Br)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2593506
View Documents
Thermo Scientific Chemicals
H2593506
5 g
Each for $144.75
Only null left
 
AAH2593514
View Documents
Thermo Scientific Chemicals
H2593514
25 g
Each for $451.92
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1996-30-1
209.442
YFFUYGSLQXVHMB-UHFFFAOYSA-N
2773264
C1=CC(=C(C=C1Cl)Br)F
C6H3BrClF
MFCD00672934
3-bromo-4-fluorochlorobenzene, 3-bromo-1-chloro-4-fluorobenzene, 2-fluoro-5-chlorobromobenzene, 1-bromo-5-chloro-2-fluorobenzene, 1-bromo-3-chloro-6-fluorobenzene, 5-chloro-2-fluorobromobenzene, 2-bromo-4-chloro-1-fluoro-benzene, 2-bromo-4-chlorofluorobenzene, benzene, 2-bromo-4-chloro-1-fluoro, pubchem3210
2-bromo-4-chloro-1-fluorobenzene

Specifications

1996-30-1
180°C
C6H3BrClF
MFCD00672934
3-bromo-4-fluorochlorobenzene, 3-bromo-1-chloro-4-fluorobenzene, 2-fluoro-5-chlorobromobenzene, 1-bromo-5-chloro-2-fluorobenzene, 1-bromo-3-chloro-6-fluorobenzene, 5-chloro-2-fluorobromobenzene, 2-bromo-4-chloro-1-fluoro-benzene, 2-bromo-4-chlorofluorobenzene, benzene, 2-bromo-4-chloro-1-fluoro, pubchem3210
C1=CC(=C(C=C1Cl)Br)F
209.442
209.45
2-Bromo-4-chloro-1-fluorobenzene
1.74
85°C (185°F)
1.554
5 g
YFFUYGSLQXVHMB-UHFFFAOYSA-N
2-bromo-4-chloro-1-fluorobenzene
2773264
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only