missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-chloro-1-fluorobenzene, 97%

Catalog No. AAH2593514
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2593514 25 g
AAH2593506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2593514 Supplier Thermo Scientific Chemicals Supplier No. H2593514
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1996-30-1 | C6H3BrClF | 209.442 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1996-30-1
Molecular Formula C6H3BrClF
Molecular Weight (g/mol) 209.442
MDL Number MFCD00672934
InChI Key YFFUYGSLQXVHMB-UHFFFAOYSA-N
Synonym 3-bromo-4-fluorochlorobenzene, 3-bromo-1-chloro-4-fluorobenzene, 2-fluoro-5-chlorobromobenzene, 1-bromo-5-chloro-2-fluorobenzene, 1-bromo-3-chloro-6-fluorobenzene, 5-chloro-2-fluorobromobenzene, 2-bromo-4-chloro-1-fluoro-benzene, 2-bromo-4-chlorofluorobenzene, benzene, 2-bromo-4-chloro-1-fluoro, pubchem3210
PubChem CID 2773264
IUPAC Name 2-bromo-4-chloro-1-fluorobenzene
SMILES C1=CC(=C(C=C1Cl)Br)F

Specifications

Density 1.74
Boiling Point 180°C
Flash Point 85°C (185°F)
Refractive Index 1.554
Quantity 25 g
Formula Weight 209.45
Percent Purity 97%
Chemical Name or Material 2-Bromo-4-chloro-1-fluorobenzene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only