missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-4,6-dinitroaniline 98.0+%, TCI America™
$40.43 - $213.69
Chemical Identifiers
| CAS | 1817-73-8 |
|---|---|
| Molecular Formula | C6H4BrN3O4 |
| Molecular Weight (g/mol) | 262.019 |
| MDL Number | MFCD00007146 |
| InChI Key | KWMDHCLJYMVBNS-UHFFFAOYSA-N |
| Synonym | 6-bromo-2,4-dinitroaniline, bromo dna, 2,4-dinitro-6-bromoaniline, benzenamine, 2-bromo-4,6-dinitro, 2-bromo-4,6-dinitroaminobenzene, 2,4-dinitro-6-bromanilin, aniline, 2,4-dinitro-6-bromo, aniline, 2-bromo-4,6-dinitro, ccris 820, 2,4-dinitro-6-bromanilin czech |
| PubChem CID | 15752 |
| IUPAC Name | 2-bromo-4,6-dinitroaniline |
| SMILES | C1=C(C=C(C(=C1[N+](=O)[O-])N)Br)[N+](=O)[O-] |
Chemical Identifiers
| 1817-73-8 | |
| 262.019 | |
| KWMDHCLJYMVBNS-UHFFFAOYSA-N | |
| 15752 | |
| C1=C(C=C(C(=C1[N+](=O)[O-])N)Br)[N+](=O)[O-] |
| C6H4BrN3O4 | |
| MFCD00007146 | |
| 6-bromo-2,4-dinitroaniline, bromo dna, 2,4-dinitro-6-bromoaniline, benzenamine, 2-bromo-4,6-dinitro, 2-bromo-4,6-dinitroaminobenzene, 2,4-dinitro-6-bromanilin, aniline, 2,4-dinitro-6-bromo, aniline, 2-bromo-4,6-dinitro, ccris 820, 2,4-dinitro-6-bromanilin czech | |
| 2-bromo-4,6-dinitroaniline |
Specifications
| 1817-73-8 | |
| C6H4BrN3O4 | |
| 25 g | |
| 6-bromo-2,4-dinitroaniline, bromo dna, 2,4-dinitro-6-bromoaniline, benzenamine, 2-bromo-4,6-dinitro, 2-bromo-4,6-dinitroaminobenzene, 2,4-dinitro-6-bromanilin, aniline, 2,4-dinitro-6-bromo, aniline, 2-bromo-4,6-dinitro, ccris 820, 2,4-dinitro-6-bromanilin czech | |
| C1=C(C=C(C(=C1[N+](=O)[O-])N)Br)[N+](=O)[O-] | |
| 262.019 | |
| 262.02 | |
| Crystalline Powder |
| 152°C | |
| MFCD00007146 | |
| 1325 | |
| KWMDHCLJYMVBNS-UHFFFAOYSA-N | |
| 2-bromo-4,6-dinitroaniline | |
| 15752 | |
| ≥98.0% (GC) | |
| 2-Bromo-4,6-dinitroaniline |
Safety and Handling
EINECSNumber : (3)-0428
RTECSNumber : BW9303000
TSCA : Yes