missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-4,6-dinitroaniline 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 152°C |
| Quantity | 500 g |
| UN Number | 1325 |
| Formula Weight | 262.02 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2-Bromo-4,6-dinitroaniline |
Safety and Handling
| EINECSNumber | (3)-0428 |
| RTECSNumber | BW9303000 |
| TSCA | Yes |