missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4,6-dimethylaniline 97.0+%, TCI America™

Catalog No. p-7119346
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B19085G 5 g
B190825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B19085G Supplier TCI America Supplier No. B19085G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 41825-73-4 | C8H10BrN | 200.08 g/mol

Chemical Identifiers

CAS 41825-73-4
Molecular Formula C8H10BrN
Molecular Weight (g/mol) 200.08
MDL Number MFCD00047826
InChI Key YOSJCQJJIHEUKA-UHFFFAOYSA-N
Synonym 2-bromo-4,6-dimethyl-aniline, 6-bromo-2,4-xylidine, benzenamine, 2-bromo-4,6-dimethyl, 2-bromo-4,6-dimethylphenylamine, 4-amino-5-bromo-m-xylene, 2-bromo-4,6-dimethylbenzenamine, 2-bromo-4,6-dimethyl-phenylamine, 2-amino-3,5-dimethyl-1-bromobenzene, pubchem3761
PubChem CID 170579
IUPAC Name 2-bromo-4,6-dimethylaniline
SMILES CC1=CC(C)=C(N)C(Br)=C1

Specifications

Melting Point 45°C
Quantity 5 g
Physical Form Crystal-Powder at 20°C
Formula Weight 200.08
Percent Purity ≥97.0% (GC,T)
Grade Not Available
Chemical Name or Material 2-Bromo-4,6-dimethylaniline