missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4,6-dimethylaniline, 98%

CAS: 41825-73-4 | C8H10BrN | 200.08 g/mol

Supplier:  Thermo Scientific Chemicals B2127209

 View more versions of this product

Catalog No. AAB2127209


Only null left
Explore available promotions

Description

Description

2-Bromo-4,6-dimethylaniline can react with 3-chloro-3-methyl-but-1-yne to produce 2-bromo-N-(1',1'-dimethylprop-2'-ynyl)-4,6-dimethylaniline. This reaction will need reagent Et3N, catalyst CuCl, and the menstruum dioxane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-4,6-dimethylaniline can react with 3-chloro-3-methyl-but-1-yne to produce 2-bromo-N-(1′,1′-dimethylprop-2′-ynyl)-4,6-dimethylaniline. This reaction will need reagent Et3N, catalyst CuCl, and the menstruum dioxane.

Solubility
Insoluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.
Specifications

Specifications

2-Bromo-4,6-dimethylaniline
41825-73-4
10 g
MFCD00047826
3046484
Insoluble in water.
CC1=CC(C)=C(N)C(Br)=C1
200.08
200.08
42°C to 46°C
75°C to 79°C (5 mmHg)
C8H10BrN
UN2811
2-bromo-4,6-dimethyl-aniline, 6-bromo-2,4-xylidine, benzenamine, 2-bromo-4,6-dimethyl, 2-bromo-4,6-dimethylphenylamine, 4-amino-5-bromo-m-xylene, 2-bromo-4,6-dimethylbenzenamine, 2-bromo-4,6-dimethyl-phenylamine, 2-amino-3,5-dimethyl-1-bromobenzene, pubchem3761
YOSJCQJJIHEUKA-UHFFFAOYSA-N
2-bromo-4,6-dimethylaniline
170579
98%

Chemical Identifiers

41825-73-4
200.08
YOSJCQJJIHEUKA-UHFFFAOYSA-N
170579
CC1=CC(C)=C(N)C(Br)=C1
C8H10BrN
MFCD00047826
2-bromo-4,6-dimethyl-aniline, 6-bromo-2,4-xylidine, benzenamine, 2-bromo-4,6-dimethyl, 2-bromo-4,6-dimethylphenylamine, 4-amino-5-bromo-m-xylene, 2-bromo-4,6-dimethylbenzenamine, 2-bromo-4,6-dimethyl-phenylamine, 2-amino-3,5-dimethyl-1-bromobenzene, pubchem3761
2-bromo-4,6-dimethylaniline
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H373-H302-H312-H315-H319
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.

P260-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P314-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H373

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 246-337-9

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only