missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4,6-dimethylaniline, 98%

Catalog No. AAB2127218
Change view
Click to view available options
Quantity:
10 g
25 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2127218 50 g
AAB2127209 10 g
AAB2127214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2127218 Supplier Thermo Scientific Chemicals Supplier No. B2127218

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 41825-73-4 | C8H10BrN | 200.08 g/mol

2-Bromo-4,6-dimethylaniline can react with 3-chloro-3-methyl-but-1-yne to produce 2-bromo-N-(1',1'-dimethylprop-2'-ynyl)-4,6-dimethylaniline. This reaction will need reagent Et3N, catalyst CuCl, and the menstruum dioxane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-4,6-dimethylaniline can react with 3-chloro-3-methyl-but-1-yne to produce 2-bromo-N-(1′,1′-dimethylprop-2′-ynyl)-4,6-dimethylaniline. This reaction will need reagent Et3N, catalyst CuCl, and the menstruum dioxane.

Solubility
Insoluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.

Chemical Identifiers

CAS 41825-73-4
Molecular Formula C8H10BrN
Molecular Weight (g/mol) 200.08
MDL Number MFCD00047826
InChI Key YOSJCQJJIHEUKA-UHFFFAOYSA-N
Synonym 2-bromo-4,6-dimethyl-aniline, 6-bromo-2,4-xylidine, benzenamine, 2-bromo-4,6-dimethyl, 2-bromo-4,6-dimethylphenylamine, 4-amino-5-bromo-m-xylene, 2-bromo-4,6-dimethylbenzenamine, 2-bromo-4,6-dimethyl-phenylamine, 2-amino-3,5-dimethyl-1-bromobenzene, pubchem3761
PubChem CID 170579
IUPAC Name 2-bromo-4,6-dimethylaniline
SMILES CC1=CC(C)=C(N)C(Br)=C1

Specifications

Melting Point 42°C to 46°C
Quantity 50 g
UN Number UN2811
Beilstein 3046484
Boiling Point 75°C to 79°C (5 mmHg)
Solubility Information Insoluble in water.
Formula Weight 200.08
Percent Purity 98%
Chemical Name or Material 2-Bromo-4,6-dimethylaniline
Hazard Category H302+H312+H332-H315-H319-H373
Hazard Statement GHS H Statement
H331-H373-H302-H312-H315-H319
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P260-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P314-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 246-337-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only