missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4,6-difluorophenol 98.0+%, TCI America™

Catalog No. B30041G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B30041G 1 g
B30045G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B30041G Supplier TCI America Supplier No. B30041G

Chemical Identifiers

CAS 98130-56-4
Molecular Formula C6H3BrF2O
Molecular Weight (g/mol) 208.99
MDL Number MFCD00070752
InChI Key YDRZYJATKMBSJX-UHFFFAOYSA-N
Synonym 2-bromo-4,6-difluoro-phenol, buttpark 20\04-78, phenol, 2-bromo-4,6-difluoro, pubchem2836, acmc-209s9n, 2-bromo-4,6-difluorophenol, 2-bromanyl-4,6-bis fluoranyl phenol, 2-bromo-4,6-difluorophenol, 98+%
PubChem CID 2736290
IUPAC Name 2-bromo-4,6-difluorophenol
SMILES C1=C(C=C(C(=C1F)O)Br)F

Specifications

Color Yellow
Boiling Point 53°C
Quantity 1 g
Formula Weight 208.99
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Bromo-4,6-difluorophenol