Learn More
2-Bromo-4,6-difluoroaniline, 98%
CAS: 444-14-4 | C6H4BrF2N | 208.01 g/mol
Supplier: Thermo Scientific Chemicals A1102222
Description
2-Bromo-4,6-difluoroaniline is used in chemical synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Bromo-4,6-difluoroaniline is used in chemical synthesis.
Solubility
Insoluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
| 2-Bromo-4,6-difluoroaniline | |
| 38°C to 42°C | |
| C6H4BrF2N | |
| 100 g | |
| 2718139 | |
| Insoluble in water. | |
| NC1=C(F)C=C(F)C=C1Br | |
| 208.01 | |
| 208.01 |
| 444-14-4 | |
| Weak, characteristic | |
| MFCD00009639 | |
| UN3077 | |
| 2,4-difluoro-6-bromoaniline, 6-bromo-2,4-difluoroaniline, 2-bromo-4,6-difluoro-phenylamine, 2-bromo-4,6-difluorobenzenamine, benzenamine, 2-bromo-4,6-difluoro, 2-bromo-4,6-difluorophenylamine, pubchem2913, acmc-1ao2h, 2-bromo4,6-difluoroaniline | |
| WUJKFVGKLTWVSQ-UHFFFAOYSA-N | |
| 2-bromo-4,6-difluoroaniline | |
| 136285 | |
| 98% |
Chemical Identifiers
| 444-14-4 | |
| 208.01 | |
| WUJKFVGKLTWVSQ-UHFFFAOYSA-N | |
| 136285 | |
| NC1=C(F)C=C(F)C=C1Br |
| C6H4BrF2N | |
| MFCD00009639 | |
| 2,4-difluoro-6-bromoaniline, 6-bromo-2,4-difluoroaniline, 2-bromo-4,6-difluoro-phenylamine, 2-bromo-4,6-difluorobenzenamine, benzenamine, 2-bromo-4,6-difluoro, 2-bromo-4,6-difluorophenylamine, pubchem2913, acmc-1ao2h, 2-bromo4,6-difluoroaniline | |
| 2-bromo-4,6-difluoroaniline |
Safety and Handling
GHS H Statement
H301
Toxic if swallowed.
P210-P233-P240-P243-P264b-P270-P280-P301+P312-P330-P374-P501c
H227-H302
DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber : 429-430-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only