missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4,6-difluoroaniline, 98%

Catalog No. AAA1102214
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1102214 25 g
AAA1102222 100 g
AAA1102236 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1102214 Supplier Thermo Scientific Chemicals Supplier No. A1102214
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 444-14-4 | C6H4BrF2N | 208.01 g/mol

2-Bromo-4,6-difluoroaniline is used in chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-4,6-difluoroaniline is used in chemical synthesis.

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 444-14-4
Molecular Formula C6H4BrF2N
Molecular Weight (g/mol) 208.01
MDL Number MFCD00009639
InChI Key WUJKFVGKLTWVSQ-UHFFFAOYSA-N
Synonym 2,4-difluoro-6-bromoaniline, 6-bromo-2,4-difluoroaniline, 2-bromo-4,6-difluoro-phenylamine, 2-bromo-4,6-difluorobenzenamine, benzenamine, 2-bromo-4,6-difluoro, 2-bromo-4,6-difluorophenylamine, pubchem2913, acmc-1ao2h, 2-bromo4,6-difluoroaniline
PubChem CID 136285
IUPAC Name 2-bromo-4,6-difluoroaniline
SMILES NC1=C(F)C=C(F)C=C1Br

Specifications

Melting Point 38°C to 42°C
Odor Weak, characteristic
Quantity 25 g
UN Number UN3077
Beilstein 2718139
Solubility Information Insoluble in water.
Formula Weight 208.01
Percent Purity 98%
Chemical Name or Material 2-Bromo-4,6-difluoroaniline
Hazard Category H227-H302
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P210-P233-P240-P243-P264b-P270-P280-P301+P312-P330-P374-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 429-430-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only