missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chemical Identifiers
| CAS | 697-86-9 |
|---|---|
| Molecular Formula | C6H4BrCl2N |
| Molecular Weight (g/mol) | 240.91 |
| MDL Number | MFCD00040936 |
| InChI Key | DTPADCOGQUOGHT-UHFFFAOYSA-N |
| Synonym | pubchem3605, acmc-209ob3, 6-bromo-2,4-dichloroaniline, 2,4-dichloro-6-bromoaniline, 2-bromo-4,6-dichloro-aniline, 6-bromo-2,4-dichlorophenylamine, benzenamine,2-bromo-4,6-dichloro, 2-bromo-4,6-dichloroaniline, 2-bromanyl-4,6-bis chloranyl aniline |
| PubChem CID | 2756901 |
| IUPAC Name | 2-bromo-4,6-dichloroaniline |
| SMILES | C1=C(C=C(C(=C1Cl)N)Br)Cl |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
AC399600050
|
N/A | 5g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| C6H4BrCl2N | |
| MFCD00040936 | |
| pubchem3605, acmc-209ob3, 6-bromo-2,4-dichloroaniline, 2,4-dichloro-6-bromoaniline, 2-bromo-4,6-dichloro-aniline, 6-bromo-2,4-dichlorophenylamine, benzenamine,2-bromo-4,6-dichloro, 2-bromo-4,6-dichloroaniline, 2-bromanyl-4,6-bis chloranyl aniline | |
| 2-bromo-4,6-dichloroaniline |
Specifications
| 697-86-9 | |
| 100.0 | |
| C6H4BrCl2N | |
| MFCD00040936 | |
| DTPADCOGQUOGHT-UHFFFAOYSA-N | |
| 2-bromo-4,6-dichloroaniline | |
| 2756901 | |
| 98% |
| 97.5 | |
| 97.5% min. (GC) | |
| Cl2C6H2(Br)NH2 | |
| pubchem3605, acmc-209ob3, 6-bromo-2,4-dichloroaniline, 2,4-dichloro-6-bromoaniline, 2-bromo-4,6-dichloro-aniline, 6-bromo-2,4-dichlorophenylamine, benzenamine,2-bromo-4,6-dichloro, 2-bromo-4,6-dichloroaniline, 2-bromanyl-4,6-bis chloranyl aniline | |
| C1=C(C=C(C(=C1Cl)N)Br)Cl | |
| 240.91 | |
| 240.91 | |
| 2-Bromo-4, 6-dichloroaniline |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a
GHS Signal Word: Warning
RUO – Research Use Only