missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4,5-dimethoxyphenol 97.0+%, TCI America™

Catalog No. b52971g
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B52971G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B52971G Supplier TCI America Supplier No. B52971G

Chemical Identifiers

CAS 129103-69-1
Molecular Formula C8H9BrO3
Molecular Weight (g/mol) 233.06
MDL Number MFCD11110624
InChI Key YGMAGXHPUIGKFF-UHFFFAOYSA-N
PubChem CID 53217002
IUPAC Name 2-bromo-4,5-dimethoxyphenol
SMILES COC1=CC(O)=C(Br)C=C1OC

Specifications

Melting Point 99°C
Quantity 1 g
Formula Weight 233.06
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-4,5-dimethoxyphenol