missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4,5-dimethoxybenzoic Acid 98.0+%, TCI America™

Catalog No. B38915G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B38915G 5 g
B38911G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B38915G Supplier TCI America Supplier No. B38915G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6286-46-0
Molecular Formula C9H9BrO4
Molecular Weight (g/mol) 261.07
MDL Number MFCD00017542
InChI Key HWFCHCRFQWEFMU-UHFFFAOYSA-N
Synonym 6-bromoveratric acid, benzoic acid, 2-bromo-4,5-dimethoxy, 2-bromo-4,5-dimethoxybenzoicacid, 6-bromo-3,4-dimethoxybenzoic acid, 6-bromveratrumsaure, acmc-1b7v8, ksc496e4l, rarechem al be 0015, timtec-bb sbb037916
PubChem CID 222963
IUPAC Name 2-bromo-4,5-dimethoxybenzoic acid
SMILES COC1=CC(Br)=C(C=C1OC)C(O)=O

Specifications

Melting Point 186°C
Color White-Yellow
Quantity 5 g
Formula Weight 261.07
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-4,5-dimethoxybenzoic Acid