missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4,5-difluorophenol, 97%

Catalog No. p-7046896
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1671603 1 g
AAL1671606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1671603 Supplier Thermo Scientific Chemicals Supplier No. L1671603
Only null left

CAS: 166281-37-4 | C6H3BrF2O | 208.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 166281-37-4
Molecular Formula C6H3BrF2O
Molecular Weight (g/mol) 208.99
MDL Number MFCD00070751
InChI Key FCYZOOHWUOEAOX-UHFFFAOYSA-N
Synonym 2-bromo-4,5-diflouo phenol, 2-bromo-4,5-difluoro-phenol, phenol, 2-bromo-4,5-difluoro, pubchem2835, acmc-1bxcg, 2-bromo-4,5-difluoro, intermediates-zcf02247, 2-bromo-4,5-difluorophen, ksc494o4b, 2-bromo-4,5-difluorophenol
PubChem CID 2736288
IUPAC Name 2-bromo-4,5-difluorophenol
SMILES OC1=CC(F)=C(F)C=C1Br

Specifications

Density 1.829
Refractive Index 1.532
Quantity 1 g
Beilstein 8355005
Formula Weight 208.99
Percent Purity 97%
Chemical Name or Material 2-Bromo-4,5-difluorophenol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only