missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4,5-difluorobenzoic acid, 97%, Thermo Scientific™

Catalog No. p-4090012
Change view
Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC399040050 5g Glass bottle
AC399040250 25g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC399040050 Supplier Thermo Scientific Chemicals Supplier No. 399040050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 64695-84-7
Molecular Formula C7H3BrF2O2
Molecular Weight (g/mol) 237
MDL Number MFCD04973422
InChI Key DGCBGBZYTNTZJH-UHFFFAOYSA-N
Synonym 2-bromo-4,5-difluorobenzoicacid, 2-bromo-4,5-difluoro-benzoic acid, benzoic acid, 2-bromo-4,5-difluoro, pubchem4991, acmc-209nnq, ksc495k3l, benzoic acid,2-bromo-4,5-difluoro, 2-bromo-4,5-difluorobenzoic acid, 2-bromanyl-4,5-bis fluoranyl benzoic acid
PubChem CID 2782789
IUPAC Name 2-bromo-4,5-difluorobenzoic acid
SMILES C1=C(C(=CC(=C1F)F)Br)C(=O)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Linear Formula F2C6H2(Br)CO2H
Quantity 5g
Formula Weight 237
Percent Purity 97%
Chemical Name or Material 2-Bromo-4, 5-difluorobenzoic acid
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only