accessibility menu, dialog, popup

UserName

FFBrOOH

CAS RN 64695-84-7

IUPAC Name: 2-bromo-4,5-difluorobenzoic acid
Synonyms: 2-bromo-4,5-difluorobenzoic acid
Molecular Weight (g/mol): 237
Molecular Formula: C7H3BrF2O2
InChi Key: DGCBGBZYTNTZJH-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(F)=C(F)C=C1Br

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)