Learn More
2-Bromo-4,5-dichloroimidazole, 98%
CAS: 16076-27-0 | C3HBrCl2N2 | 215.86 g/mol
$104.86 - $363.66
Chemical Identifiers
| CAS | 16076-27-0 |
|---|---|
| Molecular Formula | C3HBrCl2N2 |
| Molecular Weight (g/mol) | 215.86 |
| MDL Number | MFCD00067894 |
| InChI Key | SULQOTGFPPFUMG-UHFFFAOYSA-N |
| Synonym | 2-bromo-4,5-dichloroimidazole, imidazole, 2-bromo-4,5-dichloro, 1h-imidazole, 2-bromo-4,5-dichloro, imidazole,5-dichloro, acmc-20ao2s, 1h-imidazole,5-dichloro, 5-23-04-00463 beilstein handbook reference, 2-bromo-4,5-dichloro-imidazole, 2-bromo-4,5-dichloro-3h-imidazole |
| PubChem CID | 99484 |
| IUPAC Name | 2-bromo-4,5-dichloro-1H-imidazole |
| SMILES | ClC1=C(Cl)N=C(Br)N1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1489603
|
Thermo Scientific Chemicals
A1489603 |
1 g |
Each for $104.86
|
|
|||||
|
AAA1489606
|
Thermo Scientific Chemicals
A1489606 |
5 g |
Each for $363.66
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 16076-27-0 | |
| 215.86 | |
| SULQOTGFPPFUMG-UHFFFAOYSA-N | |
| 99484 | |
| ClC1=C(Cl)N=C(Br)N1 |
| C3HBrCl2N2 | |
| MFCD00067894 | |
| 2-bromo-4,5-dichloroimidazole, imidazole, 2-bromo-4,5-dichloro, 1h-imidazole, 2-bromo-4,5-dichloro, imidazole,5-dichloro, acmc-20ao2s, 1h-imidazole,5-dichloro, 5-23-04-00463 beilstein handbook reference, 2-bromo-4,5-dichloro-imidazole, 2-bromo-4,5-dichloro-3h-imidazole | |
| 2-bromo-4,5-dichloro-1H-imidazole |
Specifications
| 16076-27-0 | |
| Odorless | |
| MFCD00067894 | |
| 606732 | |
| SULQOTGFPPFUMG-UHFFFAOYSA-N | |
| 2-bromo-4,5-dichloro-1H-imidazole | |
| 99484 | |
| 98% |
| ∼215°C (decomposition) | |
| C3HBrCl2N2 | |
| 1 g | |
| 2-bromo-4,5-dichloroimidazole, imidazole, 2-bromo-4,5-dichloro, 1h-imidazole, 2-bromo-4,5-dichloro, imidazole,5-dichloro, acmc-20ao2s, 1h-imidazole,5-dichloro, 5-23-04-00463 beilstein handbook reference, 2-bromo-4,5-dichloro-imidazole, 2-bromo-4,5-dichloro-3h-imidazole | |
| ClC1=C(Cl)N=C(Br)N1 | |
| 215.86 | |
| 215.87 | |
| 2-Bromo-4,5-dichloroimidazole |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
RTECSNumber : NI3517500
TSCA : No
RUO – Research Use Only