missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4,5-dichloroimidazole, 98%

Catalog No. AAA1489603
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1489603 1 g
AAA1489606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1489603 Supplier Thermo Scientific Chemicals Supplier No. A1489603

CAS: 16076-27-0 | C3HBrCl2N2 | 215.86 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 16076-27-0
Molecular Formula C3HBrCl2N2
Molecular Weight (g/mol) 215.86
MDL Number MFCD00067894
InChI Key SULQOTGFPPFUMG-UHFFFAOYSA-N
Synonym 2-bromo-4,5-dichloroimidazole, imidazole, 2-bromo-4,5-dichloro, 1h-imidazole, 2-bromo-4,5-dichloro, imidazole,5-dichloro, acmc-20ao2s, 1h-imidazole,5-dichloro, 5-23-04-00463 beilstein handbook reference, 2-bromo-4,5-dichloro-imidazole, 2-bromo-4,5-dichloro-3h-imidazole
PubChem CID 99484
IUPAC Name 2-bromo-4,5-dichloro-1H-imidazole
SMILES ClC1=C(Cl)N=C(Br)N1

Specifications

Melting Point ∼215°C (decomposition)
Odor Odorless
Quantity 1 g
Beilstein 606732
Formula Weight 215.87
Percent Purity 98%
Chemical Name or Material 2-Bromo-4,5-dichloroimidazole
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
RTECSNumber NI3517500
TSCA No

RUO – Research Use Only