missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-3-thiophenecarboxylic acid, 96%

CAS: 24287-95-4 | C5H3BrO2S | 207.04 g/mol

$451.56 - $451.56

Chemical Identifiers

CAS 24287-95-4
Molecular Formula C5H3BrO2S
Molecular Weight (g/mol) 207.04
MDL Number MFCD05865213
InChI Key RVSXMPCELBYUSF-UHFFFAOYSA-N
Synonym 2-bromo-3-thiophenecarboxylic acid, 2-bromo-thiophene-3-carboxylic acid, 2-bromo-thiophene-3-carboxylicacid, 3-thiophenecarboxylic acid, 2-bromo, 2-bromo-3-thenoic acid, acmc-209xc0, 2-bromo-3-carboxythiophene, 2-bromothiophene-3-carboxylic?acid, 2-bromo-3-thiophene carboxylic acid, 2-bromo-thiophene 3-carboxylic acid
PubChem CID 12320971
IUPAC Name 2-bromothiophene-3-carboxylic acid
SMILES C1=CSC(=C1C(=O)O)Br
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC426840050
SDS View Documents
Thermo Scientific Chemicals
426840050
5 g Glass bottle
Each for $451.56
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

24287-95-4
207.04
RVSXMPCELBYUSF-UHFFFAOYSA-N
12320971
C1=CSC(=C1C(=O)O)Br
C5H3BrO2S
MFCD05865213
2-bromo-3-thiophenecarboxylic acid, 2-bromo-thiophene-3-carboxylic acid, 2-bromo-thiophene-3-carboxylicacid, 3-thiophenecarboxylic acid, 2-bromo, 2-bromo-3-thenoic acid, acmc-209xc0, 2-bromo-3-carboxythiophene, 2-bromothiophene-3-carboxylic?acid, 2-bromo-3-thiophene carboxylic acid, 2-bromo-thiophene 3-carboxylic acid
2-bromothiophene-3-carboxylic acid

Specifications

24287-95-4
100.0
Tan to White
96% min. (HPLC)
C5H3BrO2S
5 g
RVSXMPCELBYUSF-UHFFFAOYSA-N
2-bromothiophene-3-carboxylic acid
12320971
97%
2-Bromo-3-thiophenecarboxylic acid
96.0
179.0°C to 181.0°C
Authentic
Glass bottle
MFCD05865213
2-bromo-3-thiophenecarboxylic acid, 2-bromo-thiophene-3-carboxylic acid, 2-bromo-thiophene-3-carboxylicacid, 3-thiophenecarboxylic acid, 2-bromo, 2-bromo-3-thenoic acid, acmc-209xc0, 2-bromo-3-carboxythiophene, 2-bromothiophene-3-carboxylic?acid, 2-bromo-3-thiophene carboxylic acid, 2-bromo-thiophene 3-carboxylic acid
C1=CSC(=C1C(=O)O)Br
207.04
207.04
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only