accessibility menu, dialog, popup

UserName

CAS RN 24287-95-4

IUPAC Name: 2-bromothiophene-3-carboxylic acid
Synonyms: 2-bromothiophene-3-carboxylic acid
Molecular Weight (g/mol): 207.04
Molecular Formula: C5H3BrO2S
InChi Key: RVSXMPCELBYUSF-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(Br)SC=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results